Learn More
Poeppelmann™ KAPSTO™ GPN 270 Rectangular Plugs
Product Code. p-8003983
Click to view available options
Closure Size:
20 x 10 mm
40 x 20 mm
40 x 30 mm
50 x 20 mm
50 x 25 mm
50 x 30 mm
60 x 30 mm
60 x 40 mm
70 x 40 mm
80 x 40 mm
100 x 40 mm
100 x 50 mm
120 x 60 mm
Height (Metric):
15 mm
19 mm
22 mm
Unit Size:
Pack of 100
Pack of 25
Product Code. 10441985
Brand: Poeppelmann™ 27010400000
Only null left
Add to basket
Add to basket
Featuring distinctive external ribs to provide a secure press fit. Poeppelmann™ KAPSTO™ GPN 270 Rectangular Plugs are manufactured from black PE-LLD plastic.
  • For closing of section tubes
  • Reinforced head area for higher demands
  • High visual appeal due to grainded top surface finish and chamfered edges
Specifications
Closure Size 100 x 40 mm
Product Type Rectangular Closure Plug
For Use With (Application) For closing of profile pipes
Height (Metric) 22 mm