missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ KAPSTO™ GPN 270 Rectangular Plugs

Featuring distinctive external ribs to provide a secure press fit. Poeppelmann™ KAPSTO™ GPN 270 Rectangular Plugs are manufactured from black PE-LLD plastic.

Brand:  Poeppelmann™ 27050200000

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10023334

  • € 34.30 / Pack of 100

Estimated Shipment: 08-05-2023
to see stock.
Add to basket

Description

Description

  • For closing of section tubes
  • Reinforced head area for higher demands
  • High visual appeal due to grainded top surface finish and chamfered edges
Specifications

Specifications

50 x 20 mm
For closing of profile pipes
19 mm
Rectangular Closure Plug
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers