Learn More
Poeppelmann™ KAPSTO™ GPN 300 F Closures
Click to view available options
Diameter (Metric) Outer:
12.3 mm
14 mm
16 mm
20 mm
22 mm
25 mm
28 mm
36 mm
42 mm
48 mm
55 mm
63 mm
71 mm
95 mm
Diameter (Metric) Inner:
7.2 mm
8.4 mm
8.6 mm
9.2 mm
10.7 mm
11.7 mm
12.3 mm
13 mm
13.7 mm
14.6 mm
15.6 mm
16.5 mm
16.7 mm
17.1 mm
18.4 mm
19.3 mm
19.6 mm
21.2 mm
21.7 mm
22.4 mm
23.6 mm
24 mm
24.6 mm
26.5 mm
29 mm
30.5 mm
31 mm
32 mm
35.5 mm
36.5 mm
38.4 mm
42.8 mm
46 mm
48.5 mm
51.5 mm
55 mm
58 mm
59 mm
66 mm
86 mm
Height (Metric):
6.5 mm
7.5 mm
8 mm
8.9 mm
9 mm
10 mm
10.5 mm
10.8 mm
12 mm
12.5 mm
14.7 mm
15 mm
18 mm
21 mm
21.3 mm
21.5 mm
Unit Size:
Pack of 100
Description
- Flat-headed design
- Firm fit due to crowned shank
- Production in acc. to VDA Band 19 and ISO 16232
Specifications
Specifications
| Diameter (Metric) Outer | 20 mm |
| Product Type | Flat Top Closure Plug |
| For Use With (Application) | For internal threads and bores |
| Height (Metric) | 10 mm |
| Diameter (Metric) Inner | 15.6 mm |