Learn More
Poeppelmann™ KAPSTO™ GPN 300 F Closures
Offering multifunctional use for plugging internal threads and bore holes. Poeppelmann™ KAPSTO™ GPN 300 F Closures are manufactured from natural-colored PE-LD plastic.
Brand: Poeppelmann™ 30004020000
Additional Details : Weight : 0.50000kg
Description
- Flat-headed design
- Firm fit due to crowned shank
- Production in acc. to VDA Band 19 and ISO 16232
Specifications
Flat-headed Closure Plug | |
For internal threads and bores | |
59 mm |
63 mm | |
21.3 mm |