Learn More
Poeppelmann™ KAPSTO™ GPN 300 F Closures
Offering multifunctional use for plugging internal threads and bore holes. Poeppelmann™ KAPSTO™ GPN 300 F Closures are manufactured from natural-colored PE-LD plastic.
Brand:  Poeppelmann™ 30002330000
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10507204
  • € 13.60 / Pack of 100
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
  • Flat-headed design
  • Firm fit due to crowned shank
  • Production in acc. to VDA Band 19 and ISO 16232
Specifications
Specifications
Flat-headed Closure Plug
For internal threads and bores
26.5 mm
28 mm
12 mm