Learn More
Poeppelmann™ KAPSTO™ GPN 300 F Closures
Product Code. 10747605
Click to view available options
Diameter (Metric) Outer:
12.3 mm
14 mm
16 mm
20 mm
22 mm
25 mm
28 mm
36 mm
42 mm
48 mm
55 mm
63 mm
71 mm
95 mm
Diameter (Metric) Inner:
7.2 mm
8.4 mm
8.6 mm
9.2 mm
10.7 mm
11.7 mm
12.3 mm
13 mm
13.7 mm
14.6 mm
15.6 mm
16.5 mm
16.7 mm
17.1 mm
18.4 mm
19.3 mm
19.6 mm
21.2 mm
21.7 mm
22.4 mm
23.6 mm
24 mm
24.6 mm
26.5 mm
29 mm
30.5 mm
31 mm
32 mm
35.5 mm
36.5 mm
38.4 mm
42.8 mm
46 mm
48.5 mm
51.5 mm
55 mm
58 mm
59 mm
66 mm
86 mm
Height (Metric):
6.5 mm
7.5 mm
8 mm
8.9 mm
9 mm
10 mm
10.5 mm
10.8 mm
12 mm
12.5 mm
14.7 mm
15 mm
18 mm
21 mm
21.3 mm
21.5 mm
Unit Size:
Pack of 100
Product Code. 10747605
Brand: Poeppelmann™ 30000040000
Only null left
Add to basket
Add to basket
Offering multifunctional use for plugging internal threads and bore holes. Poeppelmann™ KAPSTO™ GPN 300 F Closures are manufactured from natural-colored PE-LD plastic.
  • Flat-headed design
  • Firm fit due to crowned shank
  • Production in acc. to VDA Band 19 and ISO 16232
Specifications
Diameter (Metric) Outer 14 mm
Product Type Flat Top Closure Plug
For Use With (Application) For internal threads and bores
Height (Metric) 6.5 mm
Diameter (Metric) Inner 9.2 mm