Learn More
Poeppelmann™ GPN 300 V Series Natural LDPE F-Type Closure Plug
Brand:  Poeppelmann™ 30100330000
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10125364
  • € 27.85 / Pack of 100
Estimated Shipment: 13-06-2024
to see stock.
Specifications
F-Type Closure Plug
48.5 mm
For recessed mounting in a bore
15 mm