Learn More
Poeppelmann™ GPN 300 V Series Natural LDPE F-Type Closure Plug
Brand: Poeppelmann™ 30100110000
Additional Details : Weight : 0.50000kg
Specifications
F-Type Closure Plug | |
14.6 mm | |
For recessed mounting in a bore | |
7.5 mm |