Learn More
Poeppelmann™ GPN 300 V Series Natural LDPE F-Type Closure Plug
Brand:  Poeppelmann™ 30101110000
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10378754
  • € 10.65 / Pack of 100
Estimated Shipment: 13-06-2024
to see stock.
Specifications
F-Type Closure Plug
15 mm
For recessed mounting in a bore
10 mm