Learn More
Poeppelmann™ GPN 350 Series Natural LDPE Protective Closure
Brand:  Poeppelmann™ 35000410000
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10207034
  • € 27.15 / Pack of 100
Estimated Shipment: 10-06-2024
to see stock.
Specifications
Protective Closure
Suitable for openings, bolts, wires and threads
12.5 H x 29.7 I.D. x 36 mm O.D.