Learn More
Poeppelmann™ GPN 350 Series Natural LDPE Protective Closure
Brand:  Poeppelmann™ 35000290000
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10461975
  • € 12.10 / Pack of 100
Estimated Shipment: 10-06-2024
to see stock.
Specifications
Protective Closure
Suitable for openings, bolts, wires and threads
12.2 H x 20.5 I.D. x 26 mm O.D.