Learn More
Poeppelmann™ GPN 550 Series Yellow LDPE Screw-in Plug
Click to view available options
Diameter (Metric) Outside:
8 mm
14 mm
18 mm
Unit Size:
Pack of 100
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | For tight closure of threads or bore holes, ideal for painting processes |
| Length (Metric) | 30 mm |
| Quantity | 100 Pack |
| Size (Metric) | 21 mm (Insertion Depth) |
| Diameter (Metric) Outside | 18 mm |
| Material | LDPE |
| Type | Screw-in Plugs |