Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 13.5mm Depth
Click to view available options
Diameter (Metric) Outside:
52 mm
54 mm
60 mm
64 mm
66 mm
Unit Size:
Pack of 100
Description
- 13.5mm insertion depth
- Universal use as plug or cap
- Tapered design ideal for accomodating tolerances
- Chemically resistant to alcohol and weak acids/alkalis
Specifications
Specifications
| Color | Natural |
| For Use With (Application) | For use as plugs in bores or internal thread, or as caps for external thread |
| Length (Metric) | 14 mm |
| Quantity | 100 Pack |
| Size (Metric) | O.D. x H: 66 x 14 mm, 13.5 mm Insertion Depth |
| Diameter (Metric) Outside | 66 mm |
| Material | LDPE |
| Type | Tapered Closure |