Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 14mm Depth
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 14mm Depth are manufactured from natural-colored PE-LD plastic.
Brand:  Poeppelmann™ 60003980000
Additional Details : Weight : 0.34200kg
 View more versions of this product
Product Code. 10227274
  • € 33.45 / Pack of 100
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • 14mm insertion depth
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Specifications
O.D. x H: 46 x 14 mm, 14 mm Insertion Depth
Tapered Closure
100 Pack
46 mm
For use as plugs in bores or internal thread, or as caps for external thread
Natural
LDPE
14 mm