Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 14mm Depth
Product Code. 10716695
Change view
Click to view available options
Diameter (Metric) Outside:
25 mm
32 mm
38 mm
40 mm
42 mm
43 mm
46 mm
47 mm
55 mm
Unit Size:
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Unit Size
10716695 55 mm Pack of 100
10748565 25 mm Pack of 100
10125264 40 mm Pack of 100
10442365 38 mm Pack of 100
10556044 32 mm Pack of 100
10767435 42 mm Pack of 100
10227274 46 mm Pack of 100
10135694 43 mm Pack of 100
10339204 47 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10716695 Supplier Poeppelmann™ Supplier No. 60005050000
  • ONLINE EXCLUSIVE PRICE
    € 50.03
    € 26.97 / Pack of 100
    Save € 23.06
    46% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 14mm Depth are manufactured from natural-colored PE-LD plastic.
  • 14mm insertion depth
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 14.2 mm
Quantity 100 Pack
Size (Metric) O.D. x H: 55 x 14.2 mm, 14 mm Insertion Depth
Diameter (Metric) Outside 55 mm
Material LDPE
Type Tapered Closure