Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 14mm Depth
Description
- 14mm insertion depth
- Universal use as plug or cap
- Tapered design ideal for accomodating tolerances
- Chemically resistant to alcohol and weak acids/alkalis
Specifications
Specifications
| Color | Natural |
| For Use With (Application) | For use as plugs in bores or internal thread, or as caps for external thread |
| Length (Metric) | 13.8 mm |
| Quantity | 100 Pack |
| Size (Metric) | O.D. x H: 25 x 13.8 mm, 14 mm Insertion Depth |
| Diameter (Metric) Outside | 25 mm |
| Material | LDPE |
| Type | Tapered Closure |