Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure (16.5mm Insertion Depth)
Product Code. 10105804
Change view
Click to view available options
Diameter (Metric) Outside:
92 mm
95 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Unit Size
10105804 95 mm Pack of 100
10093914 92 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10105804 Supplier Poeppelmann™ Supplier No. 60008400000
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 17 mm
Quantity 100 Pack
Size (Metric) O.D. x H: 95 x 17 mm, 16.5 mm Insertion Depth
Diameter (Metric) Outside 95 mm
Material LDPE
Type Tapered Closure