Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 17mm Depth
Product Code. 10287344
Change view
Click to view available options
Diameter (Metric) Outside:
88 mm
100 mm
102 mm
112 mm
115 mm
117 mm
120 mm
123 mm
Unit Size:
Pack of 100
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside unitSize
10287344 102 mm Pack of 50
10737795 112 mm Pack of 50
10747795 115 mm Pack of 50
10003894 100 mm Pack of 100
10175244 120 mm Pack of 50
10369114 88 mm Pack of 100
10013514 123 mm Pack of 50
10708175 117 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10287344 Supplier Poeppelmann™ Supplier No. 60009150000
Add to basket
Add to basket
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures are manufactured from natural-colored PE-LD plastic.
  • 17mm insertion depth
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 17.5 mm
Quantity 50 Pack
Size (Metric) O.D. x H: 102 x 17.5 mm, 17 mm Insertion Depth
Diameter (Metric) Outside 102 mm
Material LDPE
Type Tapered Closure