Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure (22.5mm Insertion Depth)
Product Code. 10537204
Change view
Click to view available options
Diameter (Metric) Outside:
209 mm
235 mm
267 mm
Unit Size:
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside unitSize
10537204 267 mm Pack of 25
10073754 209 mm Pack of 25
10083754 235 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10537204 Supplier Poeppelmann™ Supplier No. 60024900000
Add to basket
Edge
Add to basket
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 22.5 mm
Quantity 25 Pack
Size (Metric) O.D. x H: 267 x 22.5 mm, 22.5 mm Insertion Depth
Diameter (Metric) Outside 267 mm
Material LDPE
Type Tapered Closure