Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closure (22.5mm Insertion Depth)
Product Code. p-8004000
Click to view available options
Diameter (Metric) Outside:
209 mm
235 mm
267 mm
Unit Size:
Pack of 25
Product Code. 10073754
Brand: Poeppelmann™ 60019900000
Only null left
Add to basket
Edge
Add to basket
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 22.5 mm
Quantity 25 Pack
Size (Metric) O.D. x H: 209 x 22.5 mm, 22.5 mm Insertion Depth
Diameter (Metric) Outside 209 mm
Material LDPE
Type Tapered Closure