Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 23mm Depth
Product Code. 10778365
Change view
Click to view available options
Diameter (Metric) Outside:
130 mm
137.6 mm
140 mm
143 mm
149 mm
159 mm
163.6 mm
169 mm
179 mm
184 mm
213 mm
Length (Metric):
22.5 mm
23 mm
23.3 mm
23.5 mm
Unit Size:
Pack of 25
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Length (Metric) Unit Size
10778365 149 mm 23 mm Pack of 25
10083554 130 mm 23 mm Pack of 50
10175054 137.6 mm 22.5 mm Pack of 50
10237164 140 mm 23.5 mm Pack of 50
10369284 159 mm 23.3 mm Pack of 25
10338684 143 mm 22.5 mm Pack of 50
10400825 169 mm 23 mm Pack of 25
10379284 163.6 mm 23 mm Pack of 25
10034044 179 mm 23 mm Pack of 25
10165394 184 mm 22.5 mm Pack of 25
10185204 213 mm 23 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10778365 Supplier Poeppelmann™ Supplier No. 60013900000
  • ONLINE EXCLUSIVE PRICE
    € 96.82
    € 44.30 / Pack of 25
    Save € 52.52
    54% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 23mm Depth are manufactured from natural-colored PE-LD plastic.
  • 23mm insertion depth
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 23 mm
Quantity 25 Pack
Size (Metric) O.D. x H: 149 x 23 mm, 23 mm Insertion Depth
Diameter (Metric) Outside 149 mm
Material LDPE
Type Tapered Closure