Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 23mm Depth
Click to view available options
Diameter (Metric) Outside:
130 mm
137.6 mm
140 mm
143 mm
149 mm
159 mm
163.6 mm
169 mm
179 mm
184 mm
213 mm
Length (Metric):
22.5 mm
23 mm
23.3 mm
23.5 mm
Unit Size:
Pack of 25
Pack of 50
Description
- 23mm insertion depth
- Universal use as plug or cap
- Tapered design ideal for accomodating tolerances
- Chemically resistant to alcohol and weak acids/alkalis
Specifications
Specifications
| Color | Natural |
| For Use With (Application) | For use as plugs in bores or internal thread, or as caps for external thread |
| Length (Metric) | 23 mm |
| Quantity | 50 Pack |
| Size (Metric) | O.D. x H: 130 x 23 mm, 23 mm Insertion Depth |
| Diameter (Metric) Outside | 130 mm |
| Material | LDPE |
| Type | Tapered Closure |