Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 23mm Depth
Product Code. 10083554
Click to view available options
Diameter (Metric) Outside:
130 mm
137.6 mm
140 mm
143 mm
149 mm
159 mm
163.6 mm
169 mm
179 mm
184 mm
213 mm
Length (Metric):
22.5 mm
23 mm
23.3 mm
23.5 mm
Unit Size:
Pack of 25
Pack of 50
Product Code. 10083554
Brand: Poeppelmann™ 60011900000
Only null left
Add to basket
Edge
Add to basket
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 23mm Depth are manufactured from natural-colored PE-LD plastic.
  • 23mm insertion depth
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 23 mm
Quantity 50 Pack
Size (Metric) O.D. x H: 130 x 23 mm, 23 mm Insertion Depth
Diameter (Metric) Outside 130 mm
Material LDPE
Type Tapered Closure