Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures
Product Code. 10718235
Change view
Click to view available options
Diameter (Metric) Outside:
20 mm
24 mm
26 mm
28 mm
41 mm
56 mm
61 mm
72 mm
85 mm
110 mm
125 mm
159.5 mm
199.5 mm
216 mm
315 mm
Length (Metric):
8.8 mm
13 mm
13.5 mm
13.8 mm
14 mm
17 mm
17.2 mm
17.5 mm
22.5 mm
23.5 mm
33.5 mm
338 mm
Unit Size:
Pack of 100
Pack of 25
Pack of 50
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Length (Metric) Unit Size
10718235 159.5 mm 33.5 mm Pack of 25
10033654 110 mm 17.5 mm Pack of 50
10023514 125 mm 17.2 mm Pack of 50
10217764 199.5 mm 338 mm Pack of 25
10747805 20 mm 8.8 mm Pack of 100
10297404 315 mm 22.5 mm Pack of 25
10338724 41 mm 14 mm Pack of 100
10517204 26 mm 14 mm Pack of 100
10472565 216 mm 23.5 mm Pack of 25
10378714 24 mm 8.8 mm Pack of 100
10145114 28 mm 13.8 mm Pack of 100
10115364 56 mm 13.5 mm Pack of 100
10266844 61 mm 13.5 mm Pack of 100
10535485 72 mm 13 mm Pack of 100
10195314 85 mm 17 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 10718235 Supplier Poeppelmann™ Supplier No. 60014680000
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures are manufactured from natural-colored PE-LD plastic.
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 33.5 mm
Quantity 25 Pack
Size (Metric) O.D. x H: 159.5 x 33 mm
Diameter (Metric) Outside 159.5 mm
Material LDPE
Type Tapered Closure