missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poeppelmann™ GPN 600 Series Natural LDPE Tapered Closures
Click to view available options
Color:
Natural
Unit Size:
Pack of 10
Specifications
Specifications
| Diameter (Metric) | 399mm |
| For Use With (Application) | Can be used as plug or cap |
| Quantity | 10/Pk. |
| Color | Natural |
| Material | LDPE |
| Product Type | Tapered Stopper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction