Learn More
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Product Code. 10105314
Change view
Click to view available options
Diameter (Metric) Outer:
27 mm
33 mm
45 mm
48 mm
54 mm
62 mm
71 mm
83 mm
92 mm
Height (Metric):
13 mm
16 mm
19 mm
25 mm
29 mm
Unit Size:
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Height (Metric) Unit Size
10105314 27 mm 16 mm Pack of 100
10460255 45 mm 13 mm Pack of 100
10697215 33 mm 19 mm Pack of 100
10696455 48 mm 19 mm Pack of 100
10491605 83 mm 19 mm Pack of 100
10329164 92 mm 19 mm Pack of 100
10481415 71 mm 29 mm Pack of 100
10481025 62 mm 25 mm Pack of 100
10420265 54 mm 19 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10105314 Supplier Poeppelmann™ Supplier No. 61000190000
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
  • Universal use as a plug or cap
  • Acc. with VDA Band 19 and ISO 16232
  • Tapered design ideal for accomodating tolerances
Specifications
Diameter (Metric) Outer 27 mm
Product Type Universal Plug Cap
Color Yellow
Height (Metric) 16 mm
Quantity 100 Pack