Learn More
Poeppelmann™ KAPSTO™ GPN 620 Universal Protection Caps/Plugs
Featuring a tapered design for accomodating tolerances. Poeppelmann™ KAPSTO™ GPN 620 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
Brand: Poeppelmann™ 62000070000
Additional Details : Weight : 0.50000kg
Description
- Can be used as plug or cap
- For a wide range of applications
- Produced in acc. with VDA Band 19 and ISO 16232
- Same design as GPN 610 but with a larger cover flange
Specifications
Universal Protection Closure | |
For a variety of applications as a plug or cap usable | |
10 H x 10.6 I.D. x 28 mm O.D. |