Learn More
Poeppelmann™ KAPSTO™ GPN 620 Universal Protection Caps/Plugs
Product Code. 10717435
Click to view available options
Size (Metric):
10 H x 10.6 I.D. x 28 mm O.D.
11 H x 7.8 I.D. x 25 mm O.D.
12 H x 13.3 I.D. x 30 mm O.D.
13 H x 17.8 I.D. x 34 mm O.D.
15 H x 19.5 I.D. x 37 mm O.D.
15 H x 20.1 I.D. x 37 mm O.D.
19 H x 26.4 I.D. x 44 mm O.D.
19 H x 29.7 I.D. x 47 mm O.D.
19 H x 31.2 I.D. x 49 mm O.D.
Unit Size:
Pack of 100
Product Code. 10717435
Brand: Poeppelmann™ 62000030000
Only null left
Add to basket
Add to basket
Featuring a tapered design for accomodating tolerances. Poeppelmann™ KAPSTO™ GPN 620 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
  • Can be used as plug or cap
  • For a wide range of applications
  • Produced in acc. with VDA Band 19 and ISO 16232
  • Same design as GPN 610 but with a larger cover flange
Specifications
Product Type Universal Protection Closure
For Use With (Application) For a variety of applications as a plug or cap usable
Size (Metric) 11 H x 7.8 I.D. x 25 mm O.D.