missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ GPN 650 Series Yellow Form B LDPE Flange Cover Plug

Product Code. 10696635
Change view
Click to view available options
Diameter (Metric) Outside:
45 mm
68 mm
78 mm
122 mm
162 mm
205 mm
Unit Size:
Pack of 100
Pack of 25
Pack of 50
Product Code. Diameter (Metric) Outside unitSize
10696635 205 mm Pack of 25
10727245 122 mm Pack of 100
10207414 162 mm Pack of 50
10277354 45 mm Pack of 100
10185394 78 mm Pack of 100
10412735 68 mm Pack of 100
6 options
This item is not returnable. View return policy

Product Code. 10696635

Brand: Poeppelmann™ 65101250000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Yellow
For Use With (Application) For flanges according to DIN EN 1092-1 and DIN 28117
Quantity 25 Pack
Size (Metric) O.D.: 205 mm, 20 mm Insertion Depth
Diameter (Metric) Outside 205 mm
Material LDPE
Type Flange Cover Plug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.