Learn More
Poeppelmann™ KAPSTO™ GPN 650 Flange Covers
Click to view available options
Diameter (Metric) Outer:
68mm
138mm
Unit Size:
Pack of 100
Pack of 50
Description
- Acc. with DIN EN 1092-1 and DIN EN 28117
- Chemically resistant to alcohol and weak acids/alkalis
- Design B with additional clamping ribs for accomodating larger tolerances
Specifications
Specifications
| Certifications/Compliance | DIN EN 1092-1 and DIN 28117th |
| Diameter (Metric) Outer | 68mm |
| Product Type | Flange Cover Plug |