Learn More
Poeppelmann™ KAPSTO™ GPN 650 Flange Covers
Product Code. 10797425
Click to view available options
Diameter (Metric) Outer:
68mm
138mm
Unit Size:
Pack of 100
Pack of 50
Product Code. 10797425
Brand: Poeppelmann™ 65100800000
Only null left
Add to basket
Add to basket
Closes the opening and provides excellent protection for sealing faces on flange fittings. Poeppelmann™ KAPSTO™ GPN 650 Flange Covers are manufactured from yellow PE-LD plastic.
  • Acc. with DIN EN 1092-1 and DIN EN 28117
  • Chemically resistant to alcohol and weak acids/alkalis
  • Design B with additional clamping ribs for accomodating larger tolerances
Specifications
Certifications/Compliance DIN EN 1092-1 and DIN 28117th
Diameter (Metric) Outer 138mm
Product Type Flange Cover Plug