missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poeppelmann™ GPN 655 Series Yellow LDPE Flange Cover Plugs
Click to view available options
Diameter (Metric) Outside:
110 mm
Unit Size:
Pack of 100
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | Protection for flanges in accordance with ASME B 16.5 |
| Quantity | 100 Pack |
| Diameter (Metric) Outside | 110 mm |
| Material | LDPE |
| Type | Flange Cover Plug |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction