Learn More
Poeppelmann™ GPN 680 Series Yellow LDPE Flange Cap
Brand: Poeppelmann™ 68001000000
Additional Details : Weight : 0.50000kg
Specifications
Flange Caps | |
I.D. x H: 220 x 24 mm | |
24 mm | |
Suitable for fittings wit solid lining or tapered bore holes | |
Yellow |
LDPE | |
25 Pack | |
For pipe sections with flanged end, especially valves and fittings having a solid lining (e.g., lead or Rubber) or conical bores | |
220 mm |