Learn More
Poeppelmann™ GPN 680 Series Yellow LDPE Flange Cap
Product Code. 10217114
Change view
Click to view available options
Size (Metric):
I.D. x H: 115 x 19 mm
I.D. x H: 140 x 19 mm
I.D. x H: 220 x 24 mm
I.D. x H: 250 x 25 mm
I.D. x H: 285 x 25 mm
Unit Size:
Pack of 100
Pack of 25
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) unitSize
10217114 I.D. x H: 250 x 25 mm Pack of 25
10053984 I.D. x H: 220 x 24 mm Pack of 25
10207574 I.D. x H: 140 x 19 mm Pack of 50
10586825 I.D. x H: 115 x 19 mm Pack of 100
10033844 I.D. x H: 285 x 25 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10217114 Supplier Poeppelmann™ Supplier No. 68001250000
Specifications
Color Yellow
Diameter (Metric) Inside 250 mm
For Use With (Application) Suitable for fittings wit solid lining or tapered bore holes
For Use With (Equipment) For pipe sections with flanged end, especially valves and fittings having a solid lining (e.g., lead or Rubber) or conical bores
Length (Metric) 25 mm
Quantity 25 Pack
Size (Metric) I.D. x H: 250 x 25 mm
Material LDPE
Type Flange Caps