Learn More
Poeppelmann™ GPN 680 Series Yellow LDPE Flange Cap
Product Code. 10586825
Click to view available options
Size (Metric):
I.D. x H: 115 x 19 mm
I.D. x H: 140 x 19 mm
I.D. x H: 220 x 24 mm
I.D. x H: 250 x 25 mm
I.D. x H: 285 x 25 mm
Unit Size:
Pack of 100
Pack of 25
Pack of 50
Product Code. 10586825
Brand: Poeppelmann™ 68000250000
Only null left
Specifications
Color Yellow
Diameter (Metric) Inside 115 mm
For Use With (Application) Suitable for fittings wit solid lining or tapered bore holes
For Use With (Equipment) For pipe sections with flanged end, especially valves and fittings having a solid lining (e.g., lead or Rubber) or conical bores
Length (Metric) 19 mm
Quantity 100 Pack
Size (Metric) I.D. x H: 115 x 19 mm
Material LDPE
Type Flange Caps