Learn More
Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs
Click to view available options
Closure Size:
1/4 - 18 NPTF
1/4 - 28 UNF
7/16 - 20 UNF
9/16 - 18 UNF
G 5/8
M 10 x 1
M 12 x 1.5
M 16
M 20
M 20 x 2
M 22 x 1.5
M 24
M 24 x 2
M 33 x 2
M 42 x 2
M 52 x 2
M 6
Unit Size:
Pack of 100
Description
- With metric threads
- Acc. with DIN EN ISO 228-1
- Resistant to alcohol and weak acids/alkalis
- Small to medium sizes feature a slot for screwdriver installation
- Suitable for a variety of metric and imperial internal threads, as well as pipe threads
Specifications
Specifications
| Closure Size | M 52 x 2 |
| Product Type | Screw Plug Closure |
| For Use With (Application) | Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1 |