Learn More
Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs
Product Code. 10420255
Change view
Click to view available options
Closure Size:
1/4 - 18 NPTF
1/4 - 28 UNF
7/16 - 20 UNF
9/16 - 18 UNF
G 5/8
M 10 x 1
M 12 x 1.5
M 16
M 20
M 20 x 2
M 22 x 1.5
M 24
M 24 x 2
M 33 x 2
M 42 x 2
M 52 x 2
M 6
Unit Size:
Pack of 100
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size unitSize
10420255 1/4 - 28 UNF Pack of 100
10738215 M 6 Pack of 100
10748215 M 10 x 1 Pack of 100
10606825 7/16 - 20 UNF Pack of 100
10586055 M 12 x 1.5 Pack of 100
10607015 9/16 - 18 UNF Pack of 100
10318614 1/4 - 18 NPTF Pack of 100
10665495 M 16 Pack of 100
10675495 M 20 Pack of 100
10125084 M 20 x 2 Pack of 100
10758555 G 5/8 Pack of 100
10615315 M 22 x 1.5 Pack of 100
10185144 M 24 Pack of 100
10348854 M 24 x 2 Pack of 100
10527404 M 33 x 2 Pack of 100
10236984 M 42 x 2 Pack of 100
10063934 M 52 x 2 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 10420255 Supplier Poeppelmann™ Supplier No. 70072290000
Only null left
Add to basket
Add to basket
Features easy-to-grip head for manual assembly. Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs are manufactured from yellow PE-HD plastic.
  • With metric threads
  • Acc. with DIN EN ISO 228-1
  • Resistant to alcohol and weak acids/alkalis
  • Small to medium sizes feature a slot for screwdriver installation
  • Suitable for a variety of metric and imperial internal threads, as well as pipe threads
Specifications
Closure Size 1/4 - 28 UNF
Product Type Screw Plug Closure
For Use With (Application) Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1