Learn More
Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs
Product Code. p-8004014
Click to view available options
Closure Size:
1/4 - 18 NPTF
1/4 - 28 UNF
7/16 - 20 UNF
9/16 - 18 UNF
G 5/8
M 10 x 1
M 12 x 1.5
M 16
M 20
M 20 x 2
M 22 x 1.5
M 24
M 24 x 2
M 33 x 2
M 42 x 2
M 52 x 2
M 6
Unit Size:
Pack of 100
Product Code. 10420255
Brand: Poeppelmann™ 70072290000
Only null left
Add to basket
Add to basket
Features easy-to-grip head for manual assembly. Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs are manufactured from yellow PE-HD plastic.
  • With metric threads
  • Acc. with DIN EN ISO 228-1
  • Resistant to alcohol and weak acids/alkalis
  • Small to medium sizes feature a slot for screwdriver installation
  • Suitable for a variety of metric and imperial internal threads, as well as pipe threads
Specifications
Closure Size 1/4 - 28 UNF
Product Type Screw Plug Closure
For Use With (Application) Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1