Learn More
Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs
Product Code. 10368774
Change view
Click to view available options
Closure Size:
M 10
M 12
M 14
M 8 x 1
M 14 x 1.5
M 16 x 1.5
M 24 x 1.5
M 26 x 1.5
M 45 x 2
Diameter (Metric) Outer:
12 mm
14 mm
16 mm
19 mm
21 mm
31.5 mm
55 mm
Height (Metric):
12 mm
14 mm
16 mm
18 mm
20 mm
25 mm
Unit Size:
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Diameter (Metric) Outer Height (Metric) unitSize
10368774 M 8 x 1 12 mm 12 mm Pack of 100
10480255 M 10 14 mm 14 mm Pack of 100
10535674 M 12 16 mm 14 mm Pack of 100
10003674 M 14 19 mm 14 mm Pack of 100
10073674 M 16 x 1.5 21 mm 16 mm Pack of 100
10606465 M 14 x 1.5 19 mm 14 mm Pack of 100
10093434 M 24 x 1.5 31.5 mm 18 mm Pack of 100
10378924 M 26 x 1.5 31.5 mm 20 mm Pack of 100
10277394 M 45 x 2 55 mm 25 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10368774 Supplier Poeppelmann™ Supplier No. 70008100000
Add to basket
Add to basket
Features easy-to-grip head for manual assembly. Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs are manufactured from yellow PE-HD plastic.
  • With metric threads
  • Acc. with DIN EN ISO 228-1
  • Resistant to alcohol and weak acids/alkalis
  • Small to medium sizes feature a slot for screwdriver installation
  • Suitable for a variety of metric and imperial threads, as well as pipe threads
Specifications
Diameter (Metric) Outer 12 mm
Closure Size M 8 x 1
Product Type Screw Plug Closure
Color Yellow
For Use With (Application) Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1
Height (Metric) 12 mm