missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ GPN 710 Series Black Polypropylene Screw Plug

Product Code. 10013674
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10013674 Black Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10013674 Supplier Poeppelmann™ Supplier No. 71081400000
  • ONLINE EXCLUSIVE PRICE
    € 116.55
    € 82.73 / Pack of 100
    Save € 33.82
    29% Off
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Diameter (Metric) 72 mm
For Use With (Application) For installation using a spanner or screwdriver
Height (Metric) 25 mm
Quantity 100/ck.
Autoclavable Autoclavable
Color Black
Material Polypropylene
Product Type Screw Plug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.