Learn More
Poeppelmann™ KAPSTO™ GPN 720 Type A Screw Plugs
Product Code. 10013294
Click to view available options
:
Size: 16.5mm O.D.; 10 SW Metric Wrench Width
Product Code. 10013294
Brand: Poeppelmann™ 72017680000
Only null left
Add to basket
Add to basket
Features thick walls for working with higher loads. Poeppelmann™ KAPSTO™ GPN 720 Type A Screw Plugs are manufactured from yellow PE-LD plastic.
  • Immersible
  • With hexagonal socket
  • Resistant to alcohol and weak acids/alkalis
Specifications
Closure Size 1/2 - 14 NPT
Product Type Screw Plug