missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ GPN 737 Series Yellow Polyamide 6 Sealing Plug with O-ring

Product Code. 10155224
Change view
Click to view available options
Size (Metric):
22 O.D. x 21 mm H
22.2 O.D. x 17.5 mm H
27.2 O.D. x 19.5 mm H
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) unitSize
10155224 22 O.D. x 21 mm H Pack of 100
10577554 27.2 O.D. x 19.5 mm H Pack of 100
10726665 22.2 O.D. x 17.5 mm H Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10155224 Supplier Poeppelmann™ Supplier No. 73716150000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance DIN 3852-1, DIN ISO 6149-1
Diameter (Metric) Outer 22 mm
Closure Size M 16 x 1.5
Product Type Sealing Plug with O-ring
For Use With (Application) Suitable for threaded holes
Height (Metric) 21 mm
Size (Metric) 22 O.D. x 21 mm H
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.