Learn More
Poeppelmann™ KAPSTO™ GPN 800 Screw Caps
Features easy-to-grip external rib for easy mounting and removal by hand. Poeppelmann™ KAPSTO™ GPN 800 Screw Caps are manufactured from yellow PE-HD plastic.
Brand: Poeppelmann™ 80028150000
Additional Details : Weight : 0.50000kg
Description
- With metric threads
- Resistant to alcohol and weak acids/alkalis
- Suitable for a variety of different thread types
Specifications
Screw Cap | |
34.5 mm | |
100/Pk. | |
M 28 x 1.5 Threads | |
Yellow |
HDPE | |
O.D. x H: 34.5 x 30 mm, M 28 x 1.5 Thread Type, 28 mm Interior Height | |
30 mm | |
Suitable for a variety of different thread types |