Learn More
- Home
- Products
- Pumps
- Pump Parts and Accessories
- Pump Seals
- Poeppelmann™ KAPSTO™ GPN 800 Screw Caps
Poeppelmann™ KAPSTO™ GPN 800 Screw Caps
Product Code. 10687605
Change view
Click to view available options
Diameter (Metric) Outside:
12 mm
14 mm
16 mm
18 mm
21 mm
25 mm
27 mm
30 mm
32 mm
33 mm
34 mm
34.5 mm
38 mm
39 mm
40 mm
44 mm
50 mm
53 mm
55 mm
56 mm
60 mm
68 mm
Length (Metric):
9 mm
13 mm
13.5 mm
15.5 mm
16 mm
17 mm
19 mm
20 mm
21.5 mm
26 mm
30 mm
Includes:
1 5/16 - 12 Unthreaded
1 7/8 - 12 Unthreaded
7/8 - 14 UNF Threads
G 1 1/2 Threads
G 1 Threads
G 1/4 Threads
G 1/8 Threads
G 2 Threads
G 3/4 Threads
G 3/8 Threads
M 12 x 1 Threads
M 12 x 1.5 Threads
M 16 Threads
M 20 x 1.5 Threads
M 24 x 1.5 Threads
M 25 x 1.5 Threads
M 26 x 1.5 Threads
M 28 x 1.5 Threads
M 30 x 2 Threads
M 36 x 1.5 Threads
M 36 x 2 Threads
M 42 x 2 Threads
M 45 x 2 Threads
M 52 x 2 Threads
M 8 Threads
Unit Size:
Pack of 100
25 product options available for selection
| Product Code. | Diameter (Metric) Outside | Length (Metric) | Includes | unitSize |
|---|---|---|---|---|
| 10687605 | 68 mm | 20 mm | G 2 Threads | Pack of 100 |
| 10656835 | 12 mm | 9 mm | M 8 Threads | Pack of 100 |
| 10115084 | 14 mm | 9 mm | G 1/8 Threads | Pack of 100 |
| 10667415 | 16 mm | 13.5 mm | M 12 x 1.5 Threads | Pack of 100 |
| 10585874 | 16 mm | 13.5 mm | M 12 x 1 Threads | Pack of 100 |
| 10063484 | 18 mm | 13.5 mm | G 1/4 Threads | Pack of 100 |
| 10003924 | 21 mm | 13.5 mm | G 3/8 Threads | Pack of 100 |
| 10063674 | 25 mm | 15.5 mm | M 20 x 1.5 Threads | Pack of 100 |
| 10460065 | 21 mm | 13.5 mm | M 16 Threads | Pack of 100 |
| 10564715 | 30 mm | 15.5 mm | M 24 x 1.5 Threads | Pack of 100 |
| 10766845 | 32 mm | 16 mm | G 3/4 Threads | Pack of 100 |
| 10482345 | 33 mm | 16 mm | M 25 x 1.5 Threads | Pack of 100 |
| 10768415 | 34 mm | 13 mm | M 26 x 1.5 Threads | Pack of 100 |
| 10053724 | 34.5 mm | 30 mm | M 28 x 1.5 Threads | Pack of 100 |
| 10605315 | 38 mm | 16 mm | M 30 x 2 Threads | Pack of 100 |
| 10388614 | 39 mm | 16 mm | G 1 Threads | Pack of 100 |
| 10319044 | 40 mm | 26 mm | 1 5/16 - 12 Unthreaded | Pack of 100 |
| 10677415 | 27 mm | 21.5 mm | 7/8 - 14 UNF Threads | Pack of 100 |
| 10237714 | 44 mm | 19 mm | M 36 x 1.5 Threads | Pack of 100 |
| 10298234 | 44 mm | 19 mm | M 36 x 2 Threads | Pack of 100 |
| 10227214 | 56 mm | 19 mm | G 1 1/2 Threads | Pack of 100 |
| 10358924 | 53 mm | 17 mm | M 45 x 2 Threads | Pack of 100 |
| 10195144 | 55 mm | 19 mm | 1 7/8 - 12 Unthreaded | Pack of 100 |
| 10308774 | 50 mm | 17 mm | M 42 x 2 Threads | Pack of 100 |
| 10768775 | 60 mm | 17 mm | M 52 x 2 Threads | Pack of 100 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
Description
- With metric threads
- Resistant to alcohol and weak acids/alkalis
- Suitable for a variety of different thread types
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | Suitable for a variety of different thread types |
| Includes | G 2 Threads |
| Length (Metric) | 20 mm |
| Quantity | 100/Pk. |
| Size (Metric) | O.D. x H: 68 x 20 mm, G 2 Thread Type, 18 mm Interior Height |
| Diameter (Metric) Outside | 68 mm |
| Material | HDPE |
| Type | Screw Cap |