Learn More
Poeppelmann™ KAPSTO™ GPN 800 Screw Caps
Features easy-to-grip external rib for easy mounting and removal by hand. Poeppelmann™ KAPSTO™ GPN 800 Screw Caps are manufactured from yellow PE-HD plastic.
Brand: Poeppelmann™ 80003550000
Additional Details : Weight : 0.50000kg
Description
- With metric threads
- Resistant to alcohol and weak acids/alkalis
- Suitable for a variety of different thread types
Specifications
G 1/4 Threads | |
Suitable for a variety of different thread types | |
Yellow | |
HDPE | |
13.5 mm |
O.D. x H: 18 x 13.5 mm, G 1/4 Thread Type | |
Screw Cap | |
100/Pk. | |
18 mm |