Learn More
Poeppelmann™ GPN 801 Series Natural LDPE Flat Design Screw Cap
Product Code. 10083744
Click to view available options
Size (Metric):
M 22 x 1.5 Thread Type
M 3/8 Thread Type
Unit Size:
Pack of 100
Product Code. 10083744
Brand: Poeppelmann™ 80107110000
Only null left
Specifications
Color Natural
Description M 3/8 Thread Type
For Use With (Application) Suitable for outer threads
Quantity 100 Pack
Size (Metric) M 3/8 Thread Type
Material LDPE
Type Screw Cap