missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ GPN 801 Series Natural LDPE Flat Design Screw Cap

Product Code. 10287394
Click to view available options
Size (Metric):
M 22 x 1.5 Thread Type
M 3/8 Thread Type
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10287394

Brand: Poeppelmann™ 80122150000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Natural
Description M 22 x 1.5 Thread Type
For Use With (Application) Suitable for outer threads
Length (Metric) 10.5 mm
Quantity 100 Pack
Size (Metric) M 22 x 1.5 Thread Type
Diameter (Metric) Outside 25 mm
Material LDPE
Type Screw Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.