Learn More
Poeppelmann™ GPN 910 Series Black LDPE Flat Design Sealing Cover
Specifications
Specifications
| Color | Black |
| For Use With (Application) | Ideal for closing apertures in cover panels |
| Length (Metric) | 7 mm |
| Quantity | 25 Pack |
| Size (Metric) | O.D. x H: 109 x 7 mm |
| Diameter (Metric) Outside | 109 mm |
| Material | LDPE |
| Type | Sealing Cover |