Learn More
Poeppelmann™ GPN 910 Series Black LDPE Sealing Cover Plug
Product Code. 10003704
Click to view available options
Color:
Black
Unit Size:
Pack of 100
Product Code. 10003704
Brand: Poeppelmann™ 91021790000
Only null left
Specifications
Diameter (Metric) Outer 18 mm
Product Type Sealing Cover Plug
Color Black
For Use With (Application) Design for closing housing openings and blind holes
Height (Metric) 5 mm