Learn More
Poeppelmann™ GPN 985 Series LDPE Grease Nipple Cap
Brand: Poeppelmann™ 98502010000
Additional Details : Weight : 0.50000kg
Specifications
Protect the tapered head of grease nipples against contamination and visibly mark lubrication points | |
Red | |
LDPE | |
12 mm |
Grease Nipple Cap with Tab | |
100 Pack | |
12 mm |